Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Verbascoside 98%

Supplier: AMBEED, INC
Warning

A119193-1MG A119193-5MG
76797-270EA 13 USD InStock
76797-270 76797-272
Verbascoside 98%
Verbascoside
Precaución: for Research Use Only


Formula: C₂₉H₃₆O₁₅
MW: 624.59 g/mol
MDL Number: MFCD00221751
CAS Number: 61276-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty