Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-aminobutyrate hydrochloride 98%

Supplier: AMBEED, INC
Warning

A121091-100G A121091-10G A121091-25G A121091-5G
76797-366EA 62 USD InStock
76797-366 76797-368 76797-370 76797-372
Ethyl-4-aminobutyrate hydrochloride 98%
Ethyl-4-aminobutyrate hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂
MDL Number: MFCD00012915
CAS Number: 6937-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty