Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-N-methyl-D-phenylalanine 95%

Supplier: AMBEED, INC
Warning

A121129-10G A121129-1G A121129-25G A121129-5G A121129-100G
76797-374EA 43 USD InStock
76797-374 76797-376 76797-378 76797-380 76797-382 77666-738
N-Fmoc-N-methyl-D-phenylalanine 95%
N-Fmoc-N-methyl-D-phenylalanine
Precaución: for Research Use Only


Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
MDL Number: MFCD00153392
CAS Number: 138775-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty