Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(5-Bromothien-2-yl)-L-alanine 95%

Supplier: AMBEED, INC
Warning

A122009-100MG A122009-1G A122009-250MG
76797-422EA 32 USD InStock
76797-422 76797-424 76797-426
3-(5-Bromothien-2-yl)-L-alanine 95%
3-(5-Bromothien-2-yl)-L-alanine
Precaución: for Research Use Only


Formula: C₇H₈BrNO₂S
MDL Number: MFCD04117834
CAS Number: 154593-58-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty