Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-tert-Butylbenzamide 98%

Supplier: AMBEED, INC
Warning

A122144-100G A122144-10G A122144-1G A122144-25G A122144-5G
76797-428EA 430 USD InStock
76797-428 76797-430 76797-432 76797-434 76797-436
4-tert-Butylbenzamide 98%
4-tert-Butylbenzamide
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
MDL Number: MFCD00017139
CAS Number: 56108-12-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty