Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-Glyceric acid 20% in water

Supplier: AMBEED, INC
Warning

A123940-10G A123940-1G A123940-25G A123940-5G
76797-566EA 74 USD InStock
76797-566 76797-568 76797-570 76797-572
DL-Glyceric acid 20% in water
DL-Glyceric acid
Precaución: for Research Use Only


Formula: C₃H₆O₄
MW: 106.08 g/mol
MDL Number: MFCD00065927
CAS Number: 473-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty