Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Leu-Leu 95%

Supplier: AMBEED, INC
Warning

A125589-10G A125589-5G A125589-250MG A125589-1G
76797-658EA 109 USD InStock
76797-658 76797-660 77599-814 77666-792
Leu-Leu 95%
Leu-Leu
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₃
MDL Number: MFCD00065185
CAS Number: 3303-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty