Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl adipate 95%

Supplier: AMBEED, INC
Warning

A128385-1000G A128385-100G A128385-500G
76797-764EA 27 USD InStock
76797-764 76797-766 76797-768
Diethyl adipate 95%
Diethyl adipate
Precaución: for Research Use Only


Formula: C₁₀H₁₈O₄
MDL Number: MFCD00009215
CAS Number: 141-28-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty