Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Etodolac ≥99%

Supplier: AMBEED, INC
Danger

A128494-100MG A128494-1G A128494-250MG A128494-50MG A128494-5G
76797-770EA 8 USD InStock
76797-770 76797-772 76797-774 76797-776 76797-778
Etodolac ≥99%
Etodolac
Precaución: for Research Use Only


Formula: C₁₇H₂₁NO₃
MW: 287.36 g/mol
MDL Number: MFCD00133313
CAS Number: 41340-25-4
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty