Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Asp-NH₂ 98%

Supplier: AMBEED, INC
Warning

A130073-100MG A130073-250MG A130073-5G A130073-25G
76797-842EA 7 USD InStock
76797-842 76797-844 76797-846 77666-864
Fmoc-Asp-NH₂ 98%
Fmoc-Asp-NH₂
Precaución: for Research Use Only


Formula: C₁₉H₁₈N₂O₅
MDL Number: MFCD00151920
CAS Number: 200335-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty