Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Liranaftate 98%

Supplier: AMBEED, INC
Warning

A131556-100MG A131556-1G A131556-25G A131556-50MG A131556-5G
76797-892EA 7 USD InStock
76797-892 76797-894 76797-896 76797-898 76797-900
Liranaftate 98%
Liranaftate
Precaución: for Research Use Only


Formula: C₁₈H₂₀N₂O₂S
MDL Number: MFCD00865576
CAS Number: 88678-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty