Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Cys(Acm)-OH 98%

Supplier: AMBEED, INC
Warning

A131581-100G A131581-10G A131581-25G A131581-500G A131581-5G
76797-902EA 103 USD InStock
76797-902 76797-904 76797-906 76797-908 76797-910
Fmoc-Cys(Acm)-OH 98%
Fmoc-Cys(Acm)-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₂N₂O₅S
MW: 414.476 g/mol
MDL Number: MFCD00038769
CAS Number: 86060-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty