Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline 98%

Supplier: AMBEED, INC
Warning

A131713-10MG A131713-1MG A131713-5MG A131713-100MG A131713-250MG
76797-914EA 13 USD InStock
76797-914 76797-916 76797-918 77666-894 77666-896
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline 98%
4-(3-Pyridin-2-yl-1H-pyrazol-4-yl)quinoline
Precaución: for Research Use Only


Formula: C₁₇H₁₂N₄
MW: 272.31 g/mol
MDL Number: MFCD00800758
CAS Number: 396129-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty