Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1H-Indol-1-yl)propanoic acid 98%

Supplier: AMBEED, INC
Warning

A132891-1G A132891-250MG A132891-5G
76797-948EA 118 USD InStock
76797-948 76797-950 76797-952
3-(1H-Indol-1-yl)propanoic acid 98%
3-(1H-Indol-1-yl)propanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MDL Number: MFCD00022896
CAS Number: 6639-06-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty