Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mefenamic acid 95%

Supplier: AMBEED, INC
Warning

A138400-100G A138400-25G A138400-500G A138400-5G
76798-132EA 65 USD InStock
76798-132 76798-134 76798-136 76798-138
Mefenamic acid 95%
Mefenamic acid
Precaución: for Research Use Only


Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
MDL Number: MFCD00051721
CAS Number: 61-68-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty