Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-12-Ado-OH 97%

Supplier: AMBEED, INC
Warning

A140225-1G A140225-250MG A140225-25G A140225-10G
76798-212EA 20 USD InStock
76798-212 76798-214 76798-216 77667-026
Fmoc-12-Ado-OH 97%
Fmoc-12-Ado-OH
Precaución: for Research Use Only


Formula: C₂₇H₃₅NO₄
MW: 437.6 g/mol
MDL Number: MFCD00235888
CAS Number: 128917-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty