Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-1H-indole-3-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A140574-10G A140574-1G A140574-250MG A140574-25G A140574-5G
76798-224EA 119 USD InStock
76798-224 76798-226 76798-228 76798-230 76798-232
5-Methyl-1H-indole-3-carbaldehyde 98%
5-Methyl-1H-indole-3-carbaldehyde
Precaución: for Research Use Only


Formula: C10H9NO
MDL Number: MFCD00022720
CAS Number: 52562-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty