Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Cysteic acid 97%

Supplier: AMBEED, INC
Warning

A141165-100G A141165-1G A141165-250MG A141165-25G A141165-5G
76798-284EA 263 USD InStock
76798-284 76798-286 76798-288 76798-290 76798-292
L-Cysteic acid 97%
L-Cysteic acid
Precaución: for Research Use Only


Formula: C₃H₇NO₅S
MDL Number: MFCD00007524
CAS Number: 498-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty