Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chlorophenyl)acetic acid ethyl ester 95%

Supplier: AMBEED, INC
Warning

A146693-100G A146693-10G A146693-25G
76798-452EA 22 USD InStock
76798-452 76798-454 77317-120
(4-Chlorophenyl)acetic acid ethyl ester 95%
(4-Chlorophenyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: (C₆H₄Cl)CH₂COOCH₂CH₃
MW: 198.65
MDL Number: MFCD00018791
CAS Number: 14062-24-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty