Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-hydroxybutyrate 98%

Supplier: AMBEED, INC
Warning

A147392-10G A147392-100G A147392-1000G
76798-482EA 6 USD InStock
76798-482 77567-038 77667-096
Ethyl-3-hydroxybutyrate 98%
Ethyl-3-hydroxybutyrate
Precaución: For research use only.


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00004545
CAS Number: 5405-41-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty