Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-(4-aminophenyl)butanoate 97%

Supplier: AMBEED, INC
Warning

A147561-10G A147561-1G A147561-250MG A147561-5G A147561-100MG
76798-496EA 207 USD InStock
76798-496 76798-498 76798-500 76798-502 77667-098
Methyl-4-(4-aminophenyl)butanoate 97%
Methyl-4-(4-aminophenyl)butanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
MW: 193.2400 g/mol
MDL Number: MFCD07366753
CAS Number: 20637-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty