Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl pelargonate 97%

Supplier: AMBEED, INC
Warning

A149395-100G A149395-500G A149395-25G
76798-568EA 22 USD InStock
76798-568 76798-570 77299-730
Ethyl pelargonate 97%
Ethyl pelargonate
Precaución: for Research Use Only


Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
MDL Number: MFCD00009570
CAS Number: 123-29-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty