Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Thiazol-2-ylbenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A149755-100G A149755-10G A149755-1G A149755-250MG A149755-25G A149755-5G
76798-576EA 970 USD InStock
76798-576 76798-578 76798-580 76798-582 76798-584 76798-586
4-Thiazol-2-ylbenzaldehyde 97%
4-Thiazol-2-ylbenzaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₇NOS
MW: 189.24 g/mol
MDL Number: MFCD06797780
CAS Number: 198904-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty