Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethanolamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A151060-1MG A151060-500G
76798-652EA 5 USD InStock
76798-652 77667-138
Ethanolamine hydrochloride 95%
Ethanolamine hydrochloride
Precaución: For research use only.


Formula: C₂H₈ClNO
MDL Number: MFCD00012892
CAS Number: 2002-24-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty