Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lovastatin 98%

Supplier: AMBEED, INC
Warning

A151986-5G A151986-10MG A151986-500G A151986-50MG
76798-678EA 42 USD InStock
76798-678 77667-146 77667-148 77667-150
Lovastatin 98%
Lovastatin
Precaución: for Research Use Only


Formula: C₂₄H₃₆O₅
MW: 404.55 g/mol
MDL Number: MFCD00072164
CAS Number: 75330-75-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty