Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lercanidipine hydrochloride 99%

Supplier: AMBEED, INC
Danger

A152276-100MG A152276-10MG A152276-1G A152276-1MG A152276-250MG A152276-50MG A152276-5MG A152276-10G A152276-5G
76798-686EA 14 USD InStock
76798-686 76798-688 76798-690 76798-692 76798-694 76798-696 76798-698 77667-156 77667-158
Lercanidipine hydrochloride 99%
Lercanidipine hydrochloride
Precaución: for Research Use Only


Formula: C₃₆H₄₁N₃O₆·HCl
MW: 648.20 g/mol
MDL Number: MFCD07773089
CAS Number: 132866-11-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty