Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N-Trimethyldocosan-1-aminium chloride 80+%

Supplier: AMBEED, INC
Warning

A154210-1000G A154210-100G A154210-25G A154210-500G
76798-744EA 149 USD InStock
76798-744 76798-746 76798-748 76798-750
N,N,N-Trimethyldocosan-1-aminium chloride 80+%
N,N,N-Trimethyldocosan-1-aminium chloride
Precaución: for Research Use Only


Formula: C₂₅H₅₄ClN
MW: 404.15 g/mol
MDL Number: MFCD09744670
CAS Number: 17301-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty