Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ketorolac tromethamine 98%

Supplier: AMBEED, INC
Danger

A154413-100MG A154413-250MG A154413-5G
76798-752EA 5 USD InStock
76798-752 76798-754 77600-196
Ketorolac tromethamine 98%
Ketorolac tromethamine
Precaución: for Research Use Only


Formula: C₁₅H₁₃NO₃·C₄H₁₁NO₃
MDL Number: MFCD00887595
CAS Number: 74103-07-4
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty