Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium-L-aspartate hydrate 98+%

Supplier: AMBEED, INC
Warning

A154782-100G A154782-500G
76798-766EA 17 USD InStock
76798-766 77667-182
Potassium-L-aspartate hydrate 98+%
Potassium-L-aspartate hydrate
Precaución: for Research Use Only


Formula: C₄H₆KNO₄·xH₂O
MDL Number: MFCD00021083
CAS Number: 1115-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty