Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-amino-5-ureidopentanoic acid 2-hydroxysuccinic acid salt 95+%

Supplier: AMBEED, INC
Warning

A157241-100G A157241-10G A157241-1G A157241-250MG A157241-5G A157241-25G
76798-836EA 36 USD InStock
76798-836 76798-838 76798-840 76798-842 76798-844 77667-216
(S)-2-amino-5-ureidopentanoic acid 2-hydroxysuccinic acid salt 95+%
(S)-2-amino-5-ureidopentanoic acid 2-hydroxysuccinic acid salt
Precaución: for Research Use Only


Formula: C₁₀H₁₉N₃O₈
MW: 309.27 g/mol
MDL Number: MFCD09839004
CAS Number: 70796-17-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty