Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-(methylamino)-3-nitrobenzoate 98%

Supplier: AMBEED, INC
Warning

A158467-1G A158467-25G A158467-5G
76798-880EA 13 USD InStock
76798-880 76798-882 76798-884
Ethyl 4-(methylamino)-3-nitrobenzoate 98%
Ethyl 4-(methylamino)-3-nitrobenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD09746194
CAS Number: 71254-71-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty