Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6,7-dimethoxyquinoline 98%

Supplier: AMBEED, INC
Warning

A159010-100G A159010-10G A159010-25G A159010-5G A159010-1G
76798-920EA 126 USD InStock
76798-920 76798-922 76798-924 76798-926 77667-242
4-Chloro-6,7-dimethoxyquinoline 98%
4-Chloro-6,7-dimethoxyquinoline
Precaución: for Research Use Only


Formula: C₁₁H₁₀ClNO₂
MW: 223.6556 g/mol
MDL Number: MFCD07778437
CAS Number: 35654-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty