Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-methoxy-3-oxobutanoate 95%

Supplier: AMBEED, INC
Warning

A160342-10G A160342-1G
76798-984EA 83 USD InStock
76798-984 76798-986
Ethyl 4-methoxy-3-oxobutanoate 95%
Ethyl 4-methoxy-3-oxobutanoate
Precaución: for Research Use Only


Formula: C₇H₁₂O₄
MDL Number: MFCD07783018
CAS Number: 66762-68-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty