Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%

Supplier: AMBEED, INC
Warning

A161873-10G A161873-250MG A161873-1G A161873-25G
76799-026EA 1024 USD InStock
76799-026 76799-028 77592-964 77625-574
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol 97%
3-(4-Hydroxy-3-methoxyphenyl)-1-propanol
Precaución: for Research Use Only


Formula: C₁₀H₁₄O₃
MW: 182.22 g/mol
MDL Number: MFCD00016571
CAS Number: 2305-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty