Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dyclonine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A162451-1G A162451-25G A162451-50MG A162451-10MG
76799-064EA 20 USD InStock
76799-064 76799-066 76799-068 77625-582
Dyclonine hydrochloride 98%
Dyclonine hydrochloride
Precaución: for Research Use Only


Formula: C₁₈H₂₈ClNO₂
MW: 325.877 g/mol
MDL Number: MFCD00035386
CAS Number: 536-43-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty