Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Hydroquinidine 97%

Supplier: AMBEED, INC
Warning

A164149-100G A164149-100MG A164149-10G A164149-25G A164149-5G
76799-128EA 270 USD InStock
76799-128 76799-130 76799-132 76799-134 76799-136
Hydroquinidine 97%
Hydroquinidine
Precaución: for Research Use Only


Formula: C₂₀H₂₆N₂O₂
MDL Number: MFCD00135599
CAS Number: 1435-55-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty