Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine 95%

Supplier: AMBEED, INC
Warning

A167171-100MG A167171-10G A167171-1G A167171-250MG A167171-5G
76799-248EA 13 USD InStock
76799-248 76799-250 76799-252 76799-254 76799-256
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine 95%
4-(4-Chlorophenyl)-5-methyl-1,3-thiazol-2-amine
Precaución: for Research Use Only


Formula: C₁₀H₉ClN₂S
MDL Number: MFCD00731832
CAS Number: 82632-77-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty