Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(trifluoromethyl)benzoate 98%

Supplier: AMBEED, INC
Warning

A167474-100G A167474-25G A167474-5G
76799-260EA 140 USD InStock
76799-260 76799-262 77667-338
Methyl-2-(trifluoromethyl)benzoate 98%
Methyl-2-(trifluoromethyl)benzoate
Precaución: for Research Use Only


Formula: C₉H₇F₃O₂
MDL Number: MFCD00236050
CAS Number: 344-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty