Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoquinaldine 98%

Supplier: AMBEED, INC
Warning

A168290-100G A168290-1G A168290-25G A168290-500G A168290-5G
76799-296EA 243 USD InStock
76799-296 76799-298 76799-300 76799-302 76799-304
6-Bromoquinaldine 98%
6-Bromoquinaldine
Precaución: for Research Use Only


Formula: C₁₀H₈BrN
MDL Number: MFCD00079724
CAS Number: 877-42-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty