Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-amino-6-fluorobenzoate 97%

Supplier: AMBEED, INC
Warning

A168688-100G A168688-1G A168688-250MG A168688-25G A168688-5G
76799-324EA 201 USD InStock
76799-324 76799-326 76799-328 76799-330 76799-332
Methyl-2-amino-6-fluorobenzoate 97%
Methyl-2-amino-6-fluorobenzoate
Precaución: for Research Use Only


Formula: C₈H₈FNO₂
MDL Number: MFCD01763810
CAS Number: 86505-94-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty