Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diisopropyl adipate 99%

Supplier: AMBEED, INC
Warning

A169193-100G A169193-25G A169193-500G A169193-1000G
76799-338EA 27 USD InStock
76799-338 76799-340 76799-342 77625-726
Diisopropyl adipate 99%
Diisopropyl adipate
Precaución: for Research Use Only


Formula: C₁₂H₂₂O₄
MDL Number: MFCD00026391
CAS Number: 6938-94-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty