Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%

Supplier: AMBEED, INC
Warning

A170456-10G A170456-1G A170456-250MG A170456-25G A170456-5G
76799-370EA 432 USD InStock
76799-370 76799-372 76799-374 76799-376 76799-378
(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride 95%
(S)-3-Amino-3-phenylpropanoic acid ethyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
MDL Number: MFCD02259715
CAS Number: 167834-24-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty