Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl adenine-9-acetate 95%

Supplier: AMBEED, INC
Warning

A174863-10G A174863-250MG A174863-5G
76799-500EA 397 USD InStock
76799-500 76799-502 76799-504
Ethyl adenine-9-acetate 95%
Ethyl adenine-9-acetate
Precaución: for Research Use Only


Formula: C₉H₁₁N₅O₂
MW: 221.2200 g/mol
MDL Number: MFCD01321238
CAS Number: 25477-96-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty