Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropyl glycolate 95%

Supplier: AMBEED, INC
Warning

A177242-1G A177242-250MG A177242-25G A177242-5G
76799-558EA 38 USD InStock
76799-558 76799-560 76799-562 76799-564
Isopropyl glycolate 95%
Isopropyl glycolate
Precaución: for Research Use Only


Formula: HOCH₂CO₂CH(CH₃)₂
MDL Number: MFCD00046747
CAS Number: 623-61-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty