Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

JC-1 iodide salt 98%

Supplier: AMBEED, INC
Warning

A177382-1MG A177382-5MG
76799-566EA 101 USD InStock
76799-566 76799-568
JC-1 iodide salt 98%
JC-1 iodide salt
Precaución: for Research Use Only


Formula: C₂₅H₂₇Cl₄IN₄
MW: 652.23 g/mol
MDL Number: MFCD00188475
CAS Number: 47729-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty