Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ibutilide hemifumarate 98%

Supplier: AMBEED, INC
Warning

A179082-10MG A179082-1MG A179082-25MG A179082-50MG
76799-674EA 54 USD InStock
76799-674 76799-676 76799-678 76799-680
Ibutilide hemifumarate 98%
Ibutilide hemifumarate
Precaución: for Research Use Only


Formula: C₂₀H₃₆N₂O₃S
MW: 442.62 g/mol
MDL Number: MFCD01715410
CAS Number: 122647-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty