Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ecabet 98%

Supplier: AMBEED, INC
Warning

A182844-100MG A182844-1G A182844-250MG
76799-790EA 67 USD InStock
76799-790 76799-792 76799-794
Ecabet 98%
Ecabet
Precaución: for Research Use Only


Formula: C₂₀H₂₈O₅S
MDL Number: MFCD11044526
CAS Number: 33159-27-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty