Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl isobutylmalonate 98%

Supplier: AMBEED, INC
Warning

A189188-100G A189188-10G A189188-25G A189188-5G
76799-938EA 146 USD InStock
76799-938 76799-940 76799-942 77667-576
Diethyl isobutylmalonate 98%
Diethyl isobutylmalonate
Precaución: for Research Use Only


Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
MDL Number: MFCD00026869
CAS Number: 10203-58-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty