Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dihydrofluorescein diacetate 98%

Supplier: AMBEED, INC
Warning

A192744-100MG A192744-1G A192744-250MG
76800-044EA 44 USD InStock
76800-044 76800-046 76800-048
Dihydrofluorescein diacetate 98%
Dihydrofluorescein diacetate
Precaución: for Research Use Only


Formula: C₂₄H₁₈O₇
MDL Number: MFCD00005056
CAS Number: 35340-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty