Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-4-(methylamino)benzoate 98%

Supplier: AMBEED, INC
Warning

A195711-100MG A195711-1G A195711-250MG A195711-5G
76800-172EA 5 USD InStock
76800-172 76800-174 76800-176 76800-178
Ethyl-3-amino-4-(methylamino)benzoate 98%
Ethyl-3-amino-4-(methylamino)benzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O₂
MW: 194.24 g/mol
MDL Number: MFCD04110945
CAS Number: 66315-23-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty